Assay Details
Target Gene Details
Entrez Gene ID: | 228005 |
Gene Name: | peptidyl-prolyl isomerase G (cyclophilin G) |
Gene Aliases: |
AU019516, AU022200, B230312B02Rik, CYP, SRCyp |
Location: |
Chr.2:69722617-69754059 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppig | NM_001081086.1 | 14 | 2313 | NP_001074555.1 |
| AK164670.1 | 1 | 308 | BAE37869.1 | |
| BC150694.1 | 15 | 2334 |
More Information
Set Membership: |
|

Genomic Map