Assay Details
Target Gene Details
Entrez Gene ID: | 21808 |
Gene Name: | transforming growth factor, beta 2 |
Gene Aliases: |
BB105277, Tgf-beta2, Tgfb-2 |
Location: |
Chr.1:186622787-186709033 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfb2 | NM_009367.3 | 7 | 3918 | NP_033393.2 |
| AK029306.1 | 5 | 2130 | BAC26384.1 | |
| AK052377.1 | 1 | 1365 | ||
| AK084760.1 | 1 | 1194 | ||
| X57413.1 | 7 | 3917 | CAA40672.1 |
More Information
Set Membership: |
|

Genomic Map