Assay Details
Target Gene Details
Entrez Gene ID: | 73122 |
Gene Name: | transforming growth factor, beta receptor associated protein 1 |
Gene Aliases: |
3110018K12Rik, AU024090, AU042859, AW742551, Trap1 |
Location: |
Chr.1:43047265-43098622 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfbrap1 | NM_001013025.2 | 13 | 4254 | NP_001013043.1 |
| AK014063.1 | ||||
| AK141742.1 | 12 | 3810 | BAE24818.1 | |
| BC138866.1 | 13 | 4168 | ||
| BC138867.1 | 13 | 4168 | ||
| BQ959795.1 | 1 | 564 |

Genomic Map