Assay Details
Target Gene Details
Entrez Gene ID: | 12608 |
Gene Name: | CCAAT/enhancer binding protein (C/EBP), beta |
Gene Aliases: |
C/EBPbeta, CRP2, IL-6DBP, LAP, LIP, NF-IL6, NF-M, Nfil6 |
Location: |
Chr.2:167688915-167690432 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpb | NM_001287738.1 | 1 | 754 | NP_001274667.1 |
| NM_001287739.1 | 1 | 754 | NP_001274668.1 | |
| NM_009883.4 | 1 | 754 | NP_034013.1 | |
| AK143374.1 | 1 | 703 | BAE25356.1 | |
| M61007.1 | 1 | 663 | AAA37192.1 | |
| X62600.1 | 1 | 754 | CAA44484.1 |
More Information
Set Membership: |
|

Genomic Map