Assay Details
Target Gene Details
Entrez Gene ID: | 56406 |
Gene Name: | nuclear receptor coactivator 6 |
Gene Aliases: |
AIB3, ASC-2, ASC2, NRC, Ncoa7, PRIP, RAP250, mKIAA0181 |
Location: |
Chr.2:155390656-155473894 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa6 | NM_001242558.1 | 6 | 1239 | NP_001229487.1 |
| NM_019825.3 | 6 | 1239 | NP_062799.2 | |
| AF216186.1 | 5 | 1130 | AAF35860.1 | |
| AK144338.1 | 8 | 1478 | ||
| AK171864.1 | 6 | 1400 | BAE42705.1 | |
| BC031113.1 | 6 | 1243 | AAH31113.1 | |
| BC083187.1 | 6 | 1239 | AAH83187.1 | |
| JF707634.1 | 5 | 1098 | ||
| JF707635.1 | 5 | 1098 | ||
| JF707636.1 | 5 | 1098 |
More Information
Set Membership: |
|

Genomic Map