Assay Details
Target Gene Details
Entrez Gene ID: | 227731 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25 |
Gene Aliases: |
1110030N17Rik, MCSC, mKIAA1896 |
Location: |
Chr.2:32414487-32451470 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a25 | NM_001164357.1 | 10 | 3017 | NP_001157829.1 |
| NM_001164358.1 | 11 | 3272 | NP_001157830.1 | |
| NM_001290558.1 | 10 | 2886 | NP_001277487.1 | |
| NM_146118.3 | 11 | 3053 | NP_666230.2 | |
| AK003995.1 | 1 | 332 | ||
| AK049392.1 | 10 | 2993 | BAC33730.1 | |
| AK082756.1 | 10 | 2974 | BAC38604.1 | |
| AK132204.1 | 10 | 2971 | BAE21031.1 | |
| BC037109.1 | 11 | 3013 | AAH37109.1 | |
| BC066998.1 | 11 | 3240 | AAH66998.1 | |
| CO043296.1 | 1 | 307 |
More Information
Set Membership: |
|

Genomic Map