Assay Details
Target Gene Details
Entrez Gene ID: | 56695 |
Gene Name: | paroxysmal nonkinesiogenic dyskinesia |
Gene Aliases: |
2210013N15Rik, 2810403H05Rik, AI854243, Brp17, MNCb-5687, MR-1, Tahccp2 |
Location: |
Chr.1:74284597-74353692 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnkd | NM_001039509.1 | NP_001034598.1 | ||
| AK173125.1 | BAD32403.1 | |||
| BC058945.1 | AAH58945.1 |
Target Gene Details
Entrez Gene ID: | 69660 |
Gene Name: | transmembrane BAX inhibitor motif containing 1 |
Gene Aliases: |
2310061B02Rik, AA960455, AU024746, C78899, RECS1, Tmbib1, mKIAA4161 |
Location: |
Chr.1:74288247-74304336 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmbim1 | NM_027154.5 | 12 | 1895 | NP_081430.3 |
| AK077689.1 | 12 | 1720 | BAC36957.1 | |
| AK144110.1 | 12 | 1739 | BAE25704.1 | |
| AK147297.1 | 11 | 10603 | ||
| AK152864.1 | 12 | 1738 | BAE31555.1 | |
| AK166926.1 | 11 | 2137 | ||
| AK167286.1 | 7 | 1194 | BAE39392.1 | |
| AK170625.1 | 12 | 1705 | BAE41919.1 | |
| AK171102.1 | 12 | 1737 | BAE42250.1 | |
| AK179608.1 | 1 | 304 | ||
| AK220296.1 | 10 | 2901 | BAD90221.1 | |
| BC004752.1 | 5 | 1297 | AAH04752.1 | |
| BQ031095.1 | 1 | 431 |
More Information
Set Membership: |
|

Genomic Map