Assay Details
Target Gene Details
Entrez Gene ID: | 71715 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 35 |
Gene Aliases: |
1200009D07Rik, Ddx35 |
Location: |
Chr.2:158794770-158858220 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhx35 | NM_001291144.1 | 22 | 3112 | NP_001278073.1 |
| NM_145742.2 | 21 | 3020 | NP_665685.1 | |
| AK004665.1 | 22 | 3038 | ||
| AK156955.1 | 22 | 3058 | ||
| BC029709.1 | 21 | 3007 | AAH29709.1 |
More Information
Set Membership: |
|

Genomic Map