Assay Details
Target Gene Details
Entrez Gene ID: | 319909 |
Gene Name: | isthmin 1 homolog (zebrafish) |
Gene Aliases: |
5430433G21Rik, Ism |
Location: |
Chr.2:139675007-139758581 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ism1 | AK044859.1 | 1 | 296 |

Genomic Map