Assay Details
Target Gene Details
Entrez Gene ID: | 71586 |
Gene Name: | interferon induced with helicase C domain 1 |
Gene Aliases: |
9130009C22Rik, Helicard, Hlcd, MDA5, RLR-2 |
Location: |
Chr.2:62595612-62646255 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifih1 | NM_001164477.1 | 1 | 328 | NP_001157949.1 |
| NM_027835.3 | 1 | 328 | NP_082111.2 | |
| AF374384.1 | 1 | 237 | AAM21359.1 | |
| AK037057.1 | 1 | 226 | BAC29687.2 | |
| AK040519.1 | 1 | 285 | BAC30614.1 | |
| AK149802.1 | 1 | 235 | BAE29095.1 | |
| AK153018.1 | 1 | 235 | BAE31652.1 | |
| AK156450.1 | 1 | 229 | BAE33716.1 | |
| AK168646.1 | 1 | 329 | BAE40503.1 | |
| AY075132.1 | AAL77205.1 | |||
| BC080200.1 | 1 | 226 | AAH80200.1 | |
| EF113547.1 | ||||
| EF113548.1 | ||||
| EU084750.1 |

Genomic Map