Assay Details
Target Gene Details
Entrez Gene ID: | 73598 |
Gene Name: | RIKEN cDNA 1700001O22 gene |
Gene Aliases: |
1700113K14Rik, 4930549G18Rik |
Location: |
Chr.2:30795563-30803724 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700001O22Rik | NM_198000.3 | 1 | 123 | NP_932117.2 |
| NR_028486.2 | ||||
| NR_028487.2 | ||||
| AK005618.1 | ||||
| AK019754.1 | ||||
| AK076928.1 | ||||
| BC023044.1 | 1 | 93 | AAH23044.1 | |
| CX242227.1 | 1 | 123 |

Genomic Map