Assay Details
Target Gene Details
Entrez Gene ID: | 269275 |
Gene Name: | activin A receptor, type IC |
Gene Aliases: |
ACVRLK7, ALK7, Alk-7, C230097P10 |
Location: |
Chr.2:58267453-58357896 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acvr1c | NM_001033369.3 | 8 | 3597 | NP_001028541.1 |
| NM_001111030.1 | 9 | 3964 | NP_001104500.1 | |
| AK049089.1 | 8 | 3598 | BAE43339.1 | |
| AK142396.1 | 9 | 3968 |
More Information
Set Membership: |
|

Genomic Map