Assay Details
Target Gene Details
Entrez Gene ID: | 13589 |
Gene Name: | microtubule-associated protein, RP/EB family, member 1 |
Gene Aliases: |
5530600P05Rik, AI462499, AI504412, AW260097, BIM1p, D2Ertd459e, Eb1 |
Location: |
Chr.2:153741287-153773317 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapre1 | NM_007896.3 | 7 | 5181 | NP_031922.1 |
| AK017457.1 | 1 | 540 | ||
| AK046852.1 | 1 | 3158 | ||
| AK084092.1 | 1 | 3212 | ||
| AK159456.1 | 1 | 2875 |
More Information
Set Membership: |
|

Genomic Map