Assay Details
Target Gene Details
Entrez Gene ID: | 96979 |
Gene Name: | prostaglandin E synthase 2 |
Gene Aliases: |
0610038H10Rik, C79137, Gbf1, Mpges2, Pges2 |
Location: |
Chr.2:32395890-32402740 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptges2 | NM_133783.2 | 1 | 505 | NP_598544.2 |
| AK002798.1 | 1 | 130 | ||
| AK050616.1 | 1 | 131 | BAC34345.1 | |
| BC004846.1 | 1 | 137 | AAH04846.1 | |
| CX221758.1 | 1 | 505 |
More Information
Set Membership: |
|

Genomic Map