Assay Details
Target Gene Details
Entrez Gene ID: | 18108 |
Gene Name: | N-myristoyltransferase 2 |
Gene Aliases: |
A930001K02Rik, AI605445, AU044698, hNMT-2 |
Location: |
Chr.2:3284212-3328877 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nmt2 | NM_001290368.1 | 14 | 1772 | NP_001277297.1 |
| NM_001290369.1 | 13 | 1754 | NP_001277298.1 | |
| NM_001290370.1 | 12 | 1711 | NP_001277299.1 | |
| NM_008708.2 | 14 | 1804 | NP_032734.1 | |
| AF043327.1 | 14 | 1728 | AAC09297.1 | |
| AK134521.1 | 14 | 1768 | BAE22168.1 | |
| AK161490.1 | ||||
| AK168171.1 | BAE40132.1 | |||
| AK168190.1 | BAE40150.1 | |||
| BC037647.1 | AAH37647.1 |
More Information
Set Membership: |
|

Genomic Map