Assay Details
Target Gene Details
Entrez Gene ID: | 69487 |
Gene Name: | NADH dehydrogenase (ubiquinone) complex I, assembly factor 5 |
Gene Aliases: |
2310003L22Rik |
Location: |
Chr.2:140170631-140205252 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufaf5 | NM_027093.4 | 1 | 194 | NP_081369.2 |
| AK187586.1 | 1 | 60 | ||
| AK188392.1 | 1 | 60 | ||
| BC005630.1 | 1 | 61 | AAH05630.1 | |
| BY242834.1 | 1 | 195 | ||
| CD363717.1 | 1 | 164 |

Genomic Map