Assay Details
Target Gene Details
Entrez Gene ID: | 21961 |
Gene Name: | tensin 1 |
Gene Aliases: |
1110018I21Rik, 1200014E20Rik, AI648117, E030018G17Rik, E030037J05Rik, Tns |
Location: |
Chr.1:73910231-74124535 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tns1 | NM_001289895.1 | 24 | 4554 | NP_001276824.1 |
| NM_027884.3 | 26 | 4615 | NP_082160.3 | |
| AK004758.1 | 3 | 1142 | BAB23539.1 | |
| AK156648.1 | 3 | 1199 | BAE33792.1 | |
| BC052740.1 | 5 | 1380 | AAH52740.1 | |
| BC055076.1 | 3 | 1192 | AAH55076.1 |
More Information
Set Membership: |
|

Genomic Map