Assay Details
Target Gene Details
Entrez Gene ID: | 12355 |
Gene Name: | nuclear receptor subfamily 1, group I, member 3 |
Gene Aliases: |
AA209988, AI551208, CAR, CAR-beta, Care2, ESTM32, MB67 |
Location: |
Chr.1:171213935-171218845 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr1i3 | NM_001243062.1 | 8 | 1196 | NP_001229991.1 |
| NM_001243063.1 | 9 | 1299 | NP_001229992.1 | |
| NM_009803.5 | 9 | 1302 | NP_033933.2 | |
| AF009327.1 | 9 | 1239 | AAC53349.1 | |
| AF009328.1 | 8 | 1230 | AAC53350.1 | |
| AI132241.1 | 1 | 127 | ||
| AK133515.1 | 9 | 1303 | BAE21697.1 | |
| AK149381.1 | 6 | 2398 | ||
| BC037645.1 | 7 | 1372 | AAH37645.1 | |
| BC089541.1 | 9 | 1248 | AAH89541.1 |
Target Gene Details
Entrez Gene ID: | 641376 |
Gene Name: | translocase of outer mitochondrial membrane 40 homolog-like (yeast) |
Gene Aliases: |
- |
Location: |
Chr.1:171217802-171222514 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tomm40l | NM_001037170.2 | 10 | 1990 | NP_001032247.1 |
| AK028388.1 | 10 | 1991 | BAC25924.1 | |
| BC058978.1 | 9 | 1881 | AAH58978.1 |
More Information
Set Membership: |
|

Genomic Map