Assay Details
Target Gene Details
Entrez Gene ID: | 107771 |
Gene Name: | brain expressed myelocytomatosis oncogene |
Gene Aliases: |
2900002K07Rik, AW060705, Mycb |
Location: |
Chr.2:25706879-25707721 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bmyc | NM_023326.2 | 1 | 555 | NP_075815.2 |
| AK012810.1 | 1 | 597 | BAE43234.1 | |
| AK013473.1 | 1 | 605 | ||
| AK013780.1 | 1 | 602 | BAB28992.1 | |
| AK158603.1 | 1 | 605 | BAE34577.1 | |
| BC056436.1 | 1 | 555 | ||
| BC060207.1 | 1 | 555 | ||
| BC061001.1 | 1 | 654 | AAH61001.1 |
More Information
Set Membership: |
|

Genomic Map