Assay Details
Target Gene Details
Entrez Gene ID: | 16449 |
Gene Name: | jagged 1 |
Gene Aliases: |
ABE2, Gsfabe2, Htu, Ozz, Ser-1 |
Location: |
Chr.2:137081451-137116520 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jag1 | NM_013822.5 | 4 | 834 | NP_038850.1 |
| AF171092.1 | 4 | 839 | AAF15505.1 | |
| AK137092.1 | 4 | 734 | BAE23235.1 | |
| BC058675.1 | 4 | 646 | AAH58675.1 |

Genomic Map