Assay Details
Target Gene Details
Entrez Gene ID: | 381379 |
Gene Name: | mediator complex subunit 19 |
Gene Aliases: |
2410018M14Rik, 3110040A13Rik, LCMR1 |
Location: |
Chr.2:84678402-84688215 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med19 | NM_025885.3 | 1 | 43 | NP_080161.1 |
| AK005399.1 | BAC25114.1 | |||
| AK010552.1 | ||||
| AK160579.1 | BAE35886.1 | |||
| AK168787.1 | BAE40621.1 | |||
| BB611514.1 | 1 | 44 | ||
| BC082559.1 | AAH82559.1 |
Target Gene Details
Entrez Gene ID: | 66958 |
Gene Name: | thioredoxin-related transmembrane protein 2 |
Gene Aliases: |
2310042M24Rik, AA589631, Txndc14 |
Location: |
Chr.2:84671311-84678828 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmx2 | NM_025868.4 | NP_080144.1 | ||
| AK075750.1 | BAC35930.1 | |||
| BQ840236.1 |
More Information
Set Membership: |
|

Genomic Map