Assay Details
Target Gene Details
Entrez Gene ID: | 241656 |
Gene Name: | p21 protein (Cdc42/Rac)-activated kinase 7 |
Gene Aliases: |
2900083L08Rik, 6430627N20, Pak5 |
Location: |
Chr.2:136081088-136387967 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pak7 | NM_172858.2 | 1 | 264 | NP_766446.2 |
| AK013814.1 | 1 | 257 | BAE43237.1 | |
| AK032593.1 | 1 | 237 | BAC27939.1 | |
| AK077967.1 | 1 | 265 | BAE43363.1 | |
| AK079080.1 | 1 | 237 | BAC37528.1 | |
| AK163530.1 | 1 | 72 | BAE37385.1 | |
| BC099417.1 | 1 | 257 | AAH99417.1 |
More Information
Set Membership: |
|

Genomic Map