Assay Details
Target Gene Details
Entrez Gene ID: | 208263 |
Gene Name: | torsin A interacting protein 1 |
Gene Aliases: |
LAP1, Lap1B |
Location: |
Chr.1:156004599-156036480 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor1aip1 | NM_001160018.1 | 1 | 604 | NP_001153490.1 |
| NM_144791.2 | 1 | 604 | NP_659040.2 | |
| AB251963.1 | 1 | 544 | ||
| AK139709.1 | 1 | 273 | ||
| AK149727.1 | 1 | 524 | BAE29050.1 | |
| AK152751.1 | BAE31466.1 | |||
| BC010841.1 | 1 | 419 | AAH10841.1 | |
| BQ891636.1 | 1 | 199 |
Target Gene Details
Entrez Gene ID: | 240832 |
Gene Name: | torsin A interacting protein 2 |
Gene Aliases: |
1110020D10Rik, 15kDa, A130072J07, AA103493, AW060462, AW610675, C77739, Ifrg15, Lull1 |
Location: |
Chr.1:156035664-156068861 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor1aip2 | NM_001160180.1 | NP_001153652.1 | ||
| NM_172843.4 | 1 | 214 | NP_766431.3 | |
| AK038024.1 | 1 | 476 | BAC29923.1 | |
| AK150117.1 | BAE29320.1 | |||
| AK179038.1 | ||||
| BU614106.1 | 1 | 214 | ||
| BY254976.1 |

Genomic Map