Assay Details
Target Gene Details
Entrez Gene ID: | 108989 |
Gene Name: | translocated promoter region, nuclear basket protein |
Gene Aliases: |
2610029M07Rik, C77892 |
Location: |
Chr.1:150392838-150449935 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpr | NM_133780.3 | 2 | 375 | NP_598541.3 |
| AK076012.1 | 2 | 377 | BAC36119.1 | |
| AK088078.1 | 1 | 256 | BAC40132.1 | |
| AK132381.1 | 1 | 276 | BAE21135.1 | |
| AK157716.1 | 1 | 236 | BAE34164.1 | |
| BC021320.1 | 1 | 145 | ||
| BC108349.1 | 1 | 229 | AAI08350.1 | |
| BC141406.1 | 2 | 240 | ||
| BC141410.1 | 2 | 240 |
More Information
Set Membership: |
|

Genomic Map