Assay Details
Target Gene Details
Entrez Gene ID: | 170738 |
Gene Name: | potassium voltage-gated channel, subfamily H (eag-related), member 7 |
Gene Aliases: |
9330137I11Rik, Kv11.3, erg3 |
Location: |
Chr.2:62699587-63184287 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnh7 | NM_133207.2 | 4 | 752 | NP_573470.2 |
| AJ291608.1 | 4 | 559 | CAC14797.1 | |
| AK034003.1 | 4 | 753 | BAC28543.1 | |
| AK052366.1 | 4 | 636 | BAC34958.1 |
More Information
Set Membership: |
|

Genomic Map