Assay Details
Target Gene Details
Entrez Gene ID: | 77771 |
Gene Name: | cysteine-serine-rich nuclear protein 3 |
Gene Aliases: |
A330102K23Rik, CSRNP-3, Csnrp3, Mbu1, taip-2 |
Location: |
Chr.2:65845767-66031546 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csrnp3 | NM_001290665.1 | 3 | 665 | NP_001277594.1 |
| NM_001290666.1 | NP_001277595.1 | |||
| NM_153409.5 | 5 | 709 | NP_700458.3 | |
| NM_178634.2 | 4 | 547 | NP_848749.2 | |
| AB091688.1 | 2 | 351 | BAC16315.1 | |
| AK039150.1 | 5 | 635 | BAC30257.1 | |
| AK082649.1 | 4 | 472 | BAC38559.1 | |
| AK158873.1 | 3 | 667 | BAE34706.1 | |
| BC055701.1 | AAH55701.1 | |||
| EF210820.1 | 5 | 681 |

Genomic Map