Assay Details
Target Gene Details
Entrez Gene ID: | 228026 |
Gene Name: | pyruvate dehydrogenase kinase, isoenzyme 1 |
Gene Aliases: |
B830012B01, D530020C15Rik |
Location: |
Chr.2:71873218-71903858 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdk1 | NM_172665.5 | 1 | 104 | NP_766253.2 |
| AK028017.1 | ||||
| AK046805.1 | 1 | 105 | BAC32879.1 | |
| AK085215.1 | ||||
| AK153649.1 | 1 | 57 | BAE32134.1 | |
| AK163553.1 | ||||
| BC027196.1 | AAH27196.1 |
More Information
Set Membership: |
|

Genomic Map