Assay Details
Target Gene Details
Entrez Gene ID: | 209456 |
Gene Name: | transformation related protein 53 binding protein 2 |
Gene Aliases: |
53BP2, AI746547, ASPP2, PPP1R13A, Tp53bp2, X98550 |
Location: |
Chr.1:182409167-182462436 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53bp2 | NM_173378.2 | 12 | 1818 | NP_775554.2 |
| AK134556.1 | 5 | 788 | BAE22185.1 | |
| AK195318.1 | 1 | 92 | ||
| BC030894.1 | 4 | 447 | AAH30894.1 |
More Information
Set Membership: |
|

Genomic Map