Assay Details
Target Gene Details
Entrez Gene ID: | 12287 |
Gene Name: | calcium channel, voltage-dependent, N type, alpha 1B subunit |
Gene Aliases: |
AW050276, AW060892, AW822256, Cav2.2, Cchn1a, alpha(1B) |
Location: |
Chr.2:24603860-24763195 on Build GRCm38 |
Assay Gene Location: | Within Exon 48 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna1b | NM_001042528.2 | 47 | 6654 | NP_001035993.1 |
| NM_007579.3 | 46 | 6537 | NP_031605.2 | |
| AF042317.1 | 47 | 6549 | AAB97840.1 | |
| AK081230.1 | 7 | 1030 | ||
| AK134542.1 | 24 | 2972 | BAE22176.1 | |
| U04999.1 | 46 | 6552 | AAB60437.1 |
More Information
Set Membership: |
|

Genomic Map