Assay Details
Target Gene Details
Entrez Gene ID: | 13628 |
Gene Name: | eukaryotic translation elongation factor 1 alpha 2 |
Gene Aliases: |
Eef1a, S1, wasted, wst |
Location: |
Chr.2:181147692-181157015 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eef1a2 | NM_007906.2 | 4 | 850 | NP_031932.1 |
| BC018235.1 | 4 | 850 | AAH18235.1 | |
| L26479.1 | 4 | 616 | AAA91870.1 |
More Information
Set Membership: |
|

Genomic Map