Assay Details
Target Gene Details
Entrez Gene ID: | 241308 |
Gene Name: | Ral GEF with PH domain and SH3 binding motif 1 |
Gene Aliases: |
5830418G11Rik, AI853783, AI854138, RALGEF2, RALGPS1A, RalGEF 2, mKIAA0351 |
Location: |
Chr.2:33133419-33371511 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralgps1 | NM_001290570.1 | 17 | 2118 | NP_001277499.1 |
| NM_001290572.1 | 18 | 2172 | NP_001277501.1 | |
| NM_175211.5 | 20 | 2382 | NP_780420.1 | |
| AK076767.1 | 20 | 2367 | BAC36473.1 | |
| AK129123.1 | 17 | 2085 | BAC97933.1 | |
| AK170248.1 | 18 | 2120 | ||
| BC072656.1 | 18 | 2107 | AAH72656.1 |
More Information
Set Membership: |
|

Genomic Map