Assay Details
Target Gene Details
Entrez Gene ID: | 227960 |
Gene Name: | grancalcin |
Gene Aliases: |
5133401E04Rik, AI573844 |
Location: |
Chr.2:62664327-62694109 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gca | NM_145523.3 | 8 | 2851 | NP_663498.1 |
| AK050540.1 | 8 | 2893 | BAC34315.1 | |
| AK078915.1 | 8 | 2852 | BAC37457.1 | |
| AK160268.1 | 1 | 911 |

Genomic Map