Assay Details
Target Gene Details
Entrez Gene ID: | 59030 |
Gene Name: | McKusick-Kaufman syndrome |
Gene Aliases: |
1300013E18Rik, AI463362, AI957237, Bbs6 |
Location: |
Chr.2:136873780-136891505 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mkks | NM_001141946.1 | 3 | 1287 | NP_001135418.1 |
| NM_001286981.1 | 3 | 1523 | NP_001273910.1 | |
| NM_001286983.1 | NP_001273912.1 | |||
| NM_021527.2 | 3 | 1523 | NP_067502.2 | |
| AF254074.1 | 3 | 1295 | AAF73965.1 | |
| AK004987.1 | 3 | 1280 | BAB23722.1 | |
| AK032528.1 | 3 | 1285 | BAC27912.1 | |
| AK032554.1 | 3 | 1285 | BAC27921.1 | |
| BC117836.1 | 1 | 851 |
More Information
Set Membership: |
|

Genomic Map