Assay Details
Target Gene Details
Entrez Gene ID: | 12258 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade G, member 1 |
Gene Aliases: |
C1INH, C1nh |
Location: |
Chr.2:84765360-84775445 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serping1 | NM_009776.3 | 3 | 488 | NP_033906.2 |
| AF010254.1 | 3 | 463 | AAC40136.1 | |
| AK144292.1 | 2 | 449 | ||
| AK165744.1 | 2 | 436 | ||
| AK167585.1 | 2 | 426 | ||
| BC002026.1 | 2 | 419 | AAH02026.1 | |
| CA458230.1 | 3 | 525 | ||
| Y10386.1 | 3 | 445 | CAA71412.1 |
More Information
Set Membership: |
|

Genomic Map