Assay Details
Target Gene Details
Entrez Gene ID: | 107733 |
Gene Name: | mitochondrial ribosomal protein L41 |
Gene Aliases: |
MRP-L27, Rpml27 |
Location: |
Chr.2:24972470-24975098 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl41 | NM_001031808.2 | 2 | 956 | NP_001026978.2 |
| AK011837.1 | 2 | 895 | BAB27870.1 | |
| AK032370.1 | 2 | 956 | BAC27838.1 |
More Information
Set Membership: |
|

Genomic Map