Assay Details
Target Gene Details
Entrez Gene ID: | 241593 |
Gene Name: | protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, retrogene 1 |
Gene Aliases: |
OTTMUSG00000014964, Pin1-ps1, Pin1l |
Location: |
Chr.2:104713887-104716362 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pin1rt1 | NM_001033768.2 | 2 | 505 | NP_001028940.1 |
| AK145372.1 | 2 | 506 | BAE26396.1 | |
| BC147362.1 | 2 | 327 | ||
| BC147363.1 | 2 | 327 |

Genomic Map