Assay Details
Target Gene Details
Entrez Gene ID: | 12396 |
Gene Name: | core-binding factor, runt domain, alpha subunit 2, translocated to, 2 (human) |
Gene Aliases: |
A430091M07, C330013D05Rik, Cbfa2t2h, MTGR1 |
Location: |
Chr.2:154436463-154539356 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cbfa2t2 | NM_001285446.1 | 6 | 1010 | NP_001272375.1 |
| NM_009823.1 | 6 | 1014 | NP_033953.1 | |
| NM_172860.2 | 6 | 1014 | NP_766448.1 | |
| AK030597.1 | 1 | 1392 | ||
| AK040403.1 | 6 | 1006 | BAC30586.1 | |
| AK049206.1 | 6 | 972 | BAC33609.1 | |
| AK148027.1 | 6 | 1015 | BAE28297.1 | |
| AK153602.1 | 6 | 916 | BAE32114.1 | |
| BC064679.1 | 6 | 980 | AAH64679.1 | |
| BC138410.1 | 6 | 947 | ||
| BC145679.1 | 6 | 947 | ||
| DQ213025.1 | 6 | 990 | ABB02690.1 |
More Information
Set Membership: |
|

Genomic Map