Assay Details
Target Gene Details
Entrez Gene ID: | 227835 |
Gene Name: | glycosyltransferase-like domain containing 1 |
Gene Aliases: |
E330008O22Rik |
Location: |
Chr.2:44564412-44927918 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtdc1 | NM_172662.3 | 11 | 2001 | NP_766250.2 |
| AK054272.1 | 11 | 2002 | BAC35713.1 | |
| AK162289.1 | 3 | 788 | ||
| BC052069.1 | 8 | 1566 | AAH52069.1 |
More Information
Set Membership: |
|

Genomic Map