Assay Details
Target Gene Details
Entrez Gene ID: | 20612 |
Gene Name: | sialic acid binding Ig-like lectin 1, sialoadhesin |
Gene Aliases: |
Cd169, Siglec-1, Sn |
Location: |
Chr.2:131069220-131086787 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Siglec1 | NM_011426.3 | 15 | 4604 | NP_035556.3 |
| AK131121.1 | 1 | 180 | BAD21371.1 | |
| BC141335.1 | 15 | 4198 | ||
| BC141336.1 | 15 | 4194 | ||
| Z36233.1 | 15 | 4599 | CAA85268.1 | |
| Z36293.1 | 15 | 4599 | CAA85290.1 |

Genomic Map