Assay Details
Target Gene Details
Entrez Gene ID: | 329178 |
Gene Name: | unc-80 homolog (C. elegans) |
Gene Aliases: |
C030018G13Rik, C230061B10Rik, UNC-80 |
Location: |
Chr.1:66468295-66699148 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc80 | NM_175510.3 | 8 | 1108 | NP_780719.2 |
| AK044008.1 | 8 | 1141 | BAC31737.1 | |
| AK081235.1 | 8 | 1143 | BAC38173.1 | |
| FJ210934.1 | 8 | 1108 |

Genomic Map