Assay Details
Target Gene Details
Entrez Gene ID: | 269252 |
Gene Name: | general transcription factor IIIC, polypeptide 4 |
Gene Aliases: |
5330400C03, AI426938, AU014771, AU017413, KAT12, TFIIIC90 |
Location: |
Chr.2:28822299-28840360 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf3c4 | NM_001166033.1 | 6 | 3652 | NP_001159505.1 |
| NM_172977.3 | 5 | 3546 | NP_766565.2 | |
| AK030351.1 | 5 | 3473 | BAC26916.1 | |
| AK141269.1 | 5 | 3528 | ||
| AK146401.1 | 6 | 3608 | BAE27142.1 | |
| AK146403.1 | 6 | 3608 | BAE27144.1 | |
| BC061476.1 | AAH61476.1 | |||
| BC117919.1 | 5 | 3458 | ||
| BC117920.1 | 5 | 3456 |
More Information
Set Membership: |
|

Genomic Map