Assay Details
Target Gene Details
Entrez Gene ID: | 228366 |
Gene Name: | glycosyltransferase-like 1B |
Gene Aliases: |
5730485C17Rik, AI891893, Large2, Largel, mKIAA4105 |
Location: |
Chr.2:92365046-92371940 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gyltl1b | NM_001166633.2 | 11 | 1402 | NP_001160105.1 |
| NM_001290773.1 | 12 | 1510 | NP_001277702.1 | |
| NM_001290774.1 | 11 | 1424 | NP_001277703.1 | |
| NM_001290775.1 | 12 | 1727 | NP_001277704.1 | |
| NM_172670.3 | 12 | 1507 | NP_766258.2 | |
| AK077630.1 | 11 | 1378 | BAC36913.1 | |
| AK220181.1 | 3 | 3846 | BAD90366.1 | |
| AY742914.1 | 12 | 1483 | AAU95213.1 | |
| AY742915.1 | 11 | 1378 | AAU95214.1 | |
| BC033922.1 | 2 | 160 | AAH33922.1 | |
| BC096655.1 | 11 | 1434 | AAH96655.1 | |
| BC116715.1 | 11 | 1439 | ||
| HM141800.1 | 11 | 1336 |
More Information
Set Membership: |
|

Genomic Map