Assay Details
Target Gene Details
Entrez Gene ID: | 228785 |
Gene Name: | myosin, light polypeptide kinase 2, skeletal muscle |
Gene Aliases: |
9830004H17Rik, BB138278 |
Location: |
Chr.2:152911276-152923065 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mylk2 | NM_001081044.2 | 2 | 405 | NP_001074513.2 |
| AK079396.1 | 2 | 406 | ||
| BC158041.1 | 2 | 236 |
More Information
Set Membership: |
|

Genomic Map