Assay Details
Target Gene Details
Entrez Gene ID: | 19261 |
Gene Name: | signal-regulatory protein alpha |
Gene Aliases: |
AI835480, Bit, CD172a, P84, Ptpns1, SHP-1, SHPS-1, SIRP |
Location: |
Chr.2:129592606-129632228 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sirpa | NM_001177647.2 | NP_001171118.1 | ||
| NM_001291019.1 | 4 | 1373 | NP_001277948.1 | |
| NM_001291020.1 | 5 | 1175 | NP_001277949.1 | |
| NM_001291021.1 | NP_001277950.1 | |||
| NM_001291022.1 | NP_001277951.1 | |||
| NM_007547.4 | 4 | 1373 | NP_031573.2 | |
| AB018194.1 | 4 | 939 | BAA76555.1 | |
| AF072543.1 | 4 | 929 | AAC24886.1 | |
| AF072544.1 | AAC24887.1 | |||
| AF332079.1 | 4 | 939 | AAK56107.1 | |
| AF332080.1 | 4 | 939 | AAK56108.1 | |
| AK159617.1 | 5 | 1182 | ||
| AK194876.1 | 1 | 51 | ||
| BC062197.1 | 4 | 964 | AAH62197.1 | |
| CJ151068.1 | ||||
| CX236057.1 | ||||
| D85785.1 | 4 | 1223 | BAA20376.1 | |
| D87967.1 | 4 | 1338 | BAA13520.1 | |
| D87968.1 | 4 | 1338 | BAA13521.1 | |
| U89694.1 | 4 | 950 | AAB92591.1 | |
| Y10349.1 | 4 | 984 | CAA71375.1 |
More Information
Set Membership: |
|

Genomic Map