Assay Details
Target Gene Details
Entrez Gene ID: | 19261 |
Gene Name: | signal-regulatory protein alpha |
Gene Aliases: |
AI835480, Bit, CD172a, P84, Ptpns1, SHP-1, SHPS-1, SIRP |
Location: |
Chr.2:129592606-129632228 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sirpa | NM_001177647.2 | 6 | 1823 | NP_001171118.1 |
| NM_001291019.1 | 8 | 2489 | NP_001277948.1 | |
| NM_001291020.1 | 9 | 2291 | NP_001277949.1 | |
| NM_001291021.1 | 6 | 1835 | NP_001277950.1 | |
| NM_001291022.1 | 5 | 1466 | NP_001277951.1 | |
| NM_007547.4 | 8 | 2477 | NP_031573.2 | |
| AK159617.1 | 9 | 2298 | ||
| BC062197.1 | 8 | 2068 | AAH62197.1 | |
| D85785.1 | 8 | 2327 | BAA20376.1 | |
| D87967.1 | 8 | 2442 | BAA13520.1 | |
| D87968.1 | 8 | 2454 | BAA13521.1 | |
| U89694.1 | 8 | 2054 | AAB92591.1 | |
| Y10349.1 | 8 | 2100 | CAA71375.1 |
More Information
Set Membership: |
|

Genomic Map