Assay Details
Target Gene Details
Entrez Gene ID: | 22343 |
Gene Name: | lin-7 homolog C (C. elegans) |
Gene Aliases: |
9130007B12Rik, AI303698, AU019331, AW125731, D2Ertd520e, LIN-7-C, LIN-7C, MALS-3, Veli3 |
Location: |
Chr.2:109890878-109900937 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lin7c | NM_011699.3 | 5 | 2286 | NP_035829.1 |
| AK078213.1 | 1 | 1014 | ||
| AK088361.1 | 1 | 1083 | ||
| AK128989.1 | 5 | 2306 | ||
| AK161076.1 | 5 | 2267 | BAE36186.1 | |
| BC046966.1 | 5 | 2286 | AAH46966.1 |
More Information
Set Membership: |
|

Genomic Map