Assay Details
Target Gene Details
Entrez Gene ID: | 70408 |
Gene Name: | polymerase (RNA) III (DNA directed) polypeptide F |
Gene Aliases: |
2810411G20Rik, 3010019O03Rik, 3110032A07Rik, RPC39, RPC6 |
Location: |
Chr.2:144527745-144541779 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr3f | NM_029763.3 | 9 | 2567 | NP_084039.2 |
| AK029286.1 | 9 | 2568 | BAC26372.1 | |
| AK036166.1 | 9 | 2564 | BAC29327.1 | |
| AK144679.1 | 9 | 2565 | BAE26008.1 |
More Information
Set Membership: |
|

Genomic Map