Assay Details
Target Gene Details
Entrez Gene ID: | 11305 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 2 |
Gene Aliases: |
AI413825, Abc2, D2H0S1474E, mKIAA1062 |
Location: |
Chr.2:25428604-25448544 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca2 | NM_007379.2 | 8 | 1108 | NP_031405.2 |
| AK169438.1 | 8 | 1108 | ||
| X75927.2 | 8 | 1012 | CAA53531.2 |
More Information
Set Membership: |
|

Genomic Map