Assay Details
Target Gene Details
Entrez Gene ID: | 229003 |
Gene Name: | helicase with zinc finger 2, transcriptional coactivator |
Gene Aliases: |
PDIP1, Pric285, mKIAA1769, mPDIP1 |
Location: |
Chr.2:181227615-181242058 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Helz2 | NM_183162.2 | 2 | 428 | NP_898985.2 |
| AB201714.1 | 1 | 332 | ||
| AK157942.1 | 2 | 427 | BAE34274.1 | |
| AK168622.1 | 2 | 429 | ||
| AK172227.1 | 2 | 519 | BAE42894.1 |
More Information
Set Membership: |
|

Genomic Map