Assay Details
Target Gene Details
Entrez Gene ID: | 21749 |
Gene Name: | telomeric repeat binding factor 1 |
Gene Aliases: |
Pin2, Trbf1, Trf1 |
Location: |
Chr.1:15804982-15844052 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Terf1 | NM_001286628.1 | 9 | 1213 | NP_001273557.1 |
| NM_009352.3 | 10 | 1300 | NP_033378.1 | |
| AB109563.1 | 10 | 1280 | BAC76782.1 | |
| AK132022.1 | 9 | 1196 | BAE20947.1 | |
| BC034866.1 | 10 | 1288 | AAH34866.1 | |
| U65586.1 | 10 | 1280 | AAB53970.1 |

Genomic Map